Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Ideal for use in a rotation program with LpH™ se One-Step Germicidal Disinfectant (14-415-10). Suitable for use in institutions such as pharmaceutical plants, veterinary clinics and industrial plants.
- Contains phenols blended with synthetic detergents and builders
- Kills a broad spectrum of common bacteria at a use dilution rate of 1:128 (1 oz. per gal. [30mL per 3.8L]) without harming surfaces
- Also effective against fungi and viruses, including the HIV-1 virus, Herpes Simplex Type 2, Influenza A2 (Japan)
- Effective with soft water or hard water up to 400ppm hardness (as CaCO3) and in the presence of 5% serum
- Advanced detergent system provides superior cleaning and disinfecting
Specifications
Specifications
| pH | 12.3 |
| Appearance | Red |
| Certifications/Compliance | NOISH, OSHA |
| Composition | 9.09% 2-Phenylphenol, 7.66% p-Tertiary Amylphenol, 5% Potassium Hydroxide, <2% Sodium Hydroxide |
| For Use With (Application) | Disinfecting blood spills in institutions such as hospitals, nursing homes, schools, medical and dental offices, pharmaceutical plants and other indoor areas where disinfection, cleaning and deodorizing are necessary |
| Volume (English) | 1 gal. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?