Learn More
MilliporeSigma STERITEST CANNISTER (BLUE)
Catalog No. NC9314162
Encompass
Steritest EZ Device, For liquids in ampoules or collapsible bags, Blue, Styrene acrylonitrile, Filter Material: Mixed cellulose esters, 0.45um, 120mL, Large volume parenterals and synthetic drugs without antimicrobial activity, 45deg.C
Catalog No. NC9314162 Supplier MilliporeSigma Supplier No. TZHALA210
Steritest EZ Device, For liquids in ampoules or collapsible bags, Blue, Styrene acrylonitrile, Filter Material: Mixed cellulose esters, 0.45um, 120mL, Large volume parenterals and synthetic drugs without antimicrobial activity, 45deg.C