Learn More
MilliporeSigma STERITEST CANNISTER (RED)
Supplier:  MilliporeSigma TZHVAB210
Encompass
Steritest EZ Device, For antibiotics, Red, Hydrophilic PVDF, 0.45um, 120mL, Antibiotics and products containing antimicrobial agents, 45deg.C
Catalog No. NC9313965