Learn More
MilliporeSigma Steritest NEO Device
Catalog No. NC0929367
Encompass
Steritest NEO is a membrane filtration device for sterility testing of filterable pharmaceutical products. The device simplifies every aspect of testing, from handling to traceability. The closed system ensures that pharmaceutical products are never exposed to the environment, minimizes false positives, and offers the highest levels of quality and reliability.
Catalog No. NC0929367 Supplier MilliporeSigma Supplier No. TZHVMD210
Steritest NEO is a membrane filtration device for sterility testing of filterable pharmaceutical products. The device simplifies every aspect of testing, from handling to traceability. The closed system ensures that pharmaceutical products are never exposed to the environment, minimizes false positives, and offers the highest levels of quality and reliability.