Learn More
MilliporeSigma STERITEST KITS 10/PKSTERITEST
Catalog No. NC9303422
Encompass
Steritest EZ Device, For liquids in vials, Blue, Styrene acrylonitrile, Filter Material: Mixed cellulose esters, 0.45um, 120mL, Large volume parenterals and synthetic drugs without antimicrobial activity, 45deg.C
Catalog No. NC9303422 Supplier MilliporeSigma Supplier No. TZHALV210
Steritest EZ Device, For liquids in vials, Blue, Styrene acrylonitrile, Filter Material: Mixed cellulose esters, 0.45um, 120mL, Large volume parenterals and synthetic drugs without antimicrobial activity, 45deg.C