Learn More
Thomas Scientific STERIWARE BLUE LIQUID FUNNEL,
SDP
Catalog No. NC1733292
Encompass
SteriWare Blue Liquid Funnel, PP,100mm Top Diameter, Sterile
Catalog No. NC1733292 Supplier Thomas Scientific Supplier No. 1140H74
SteriWare Blue Liquid Funnel, PP,100mm Top Diameter, Sterile