Learn More
Health Care Logistics Inc Sterile Empty Vials, Clear, 5mL, 25/pk
SDP
Catalog No. NC1665921
Encompass
5mL empty sterile glass vials are dry heat sterilized and individually inspected. Sterile vials have 20mm caps. Six sizes (2mL, 5mL, 10mL, 30mL, 50mL, 100mL) provide variety to meet a range of medication preparation demands. Stoppers are latex free. EP Type 1 borosilicate glass. Empty Sterile Glass Vials are pyrogen free.
This item is not returnable. View return policy
Catalog No. NC1665921 Supplier Health Care Logistics Inc Supplier No. 18492
5mL empty sterile glass vials are dry heat sterilized and individually inspected. Sterile vials have 20mm caps. Six sizes (2mL, 5mL, 10mL, 30mL, 50mL, 100mL) provide variety to meet a range of medication preparation demands. Stoppers are latex free. EP Type 1 borosilicate glass. Empty Sterile Glass Vials are pyrogen free.