Learn More
CPC Scientific Biotin-Cys-Pro-Ser-Ser-His-Ser-Ser-Leu-Thr-Glu-Arg-His-Lys-Ile-Leu-His-Arg-Leu-Leu-Gln-Glu-Gly-Ser-Pro-Ser-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110411
Encompass
This is amino acids 676 to 700 fragment of steroid receptor co-activator (SRC1). This N-terminnaly biotinylated peptide is derived from the second LXXLL motif of SRC1. Co-activator proteins interact with nuclear receptors in a ligand-dependent manner and augment transcription. A short amphipathic betalpha-Helical domain that includes this LXXLL motif serves as the interaction interface between the co-activator molecules and the ligand-dependent activation function (AF-2) located in the COOH-terminus of the nuclear receptor ligand-binding domain (LBD). Receptor binding domain of the co-activator SRC-1 is specifically recruited to the farnesoid X receptor (FXR) ligand-binding domainONE-LETTER SEQUENCE: Biotin-CPSSHSSLTERHKILHRLLQEGSPSMOLECULAR FORMULA: C128H209N41O40S2MOLECULAR WEIGHT:3026.42STORAGE CONDITIONS: -20 5CRESEARCH AREA: Inflammation
Catalog No. 50-211-0411 Supplier CPC Scientific Supplier No. SIGN015A
This is amino acids 676 to 700 fragment of steroid receptor co-activator (SRC1). This N-terminnaly biotinylated peptide is derived from the second LXXLL motif of SRC1. Co-activator proteins interact with nuclear receptors in a ligand-dependent manner and augment transcription. A short amphipathic betalpha-Helical domain that includes this LXXLL motif serves as the interaction interface between the co-activator molecules and the ligand-dependent activation function (AF-2) located in the COOH-terminus of the nuclear receptor ligand-binding domain (LBD). Receptor binding domain of the co-activator SRC-1 is specifically recruited to the farnesoid X receptor (FXR) ligand-binding domainONE-LETTER SEQUENCE: Biotin-CPSSHSSLTERHKILHRLLQEGSPSMOLECULAR FORMULA: C128H209N41O40S2MOLECULAR WEIGHT:3026.42STORAGE CONDITIONS: -20 5CRESEARCH AREA: Inflammation