Learn More
Cayman Chemical Steryl GlucosIdes 10mg
SDP
Catalog No. 502939950
Encompass_Preferred
A mixture of steryl glucosides; neutral glycolipids commonly found in plant cell membranes and vegetable oils; glucose donors in the biosynthesis of glucocerebrosides; metabolic precursors to acylated/esterified steryl glucosides; major component of filter- and engine-damaging precipitates formed during biodiesel production from transesterification of vegetable oils
Catalog No. 50-293-9950 Supplier Cayman Chemical Supplier No. 2720510 MG
A mixture of steryl glucosides; neutral glycolipids commonly found in plant cell membranes and vegetable oils; glucose donors in the biosynthesis of glucocerebrosides; metabolic precursors to acylated/esterified steryl glucosides; major component of filter- and engine-damaging precipitates formed during biodiesel production from transesterification of vegetable oils