Learn More
Bioworld STEBuffer (1X), 4 L
Catalog No. 50196670
Encompass_Preferred
STE Reagent Common reagent for molecular biology. Used in DNA extraction, DNA electrophoresis and many other lab protocols.DNAse and RNAse free, ready to use 1X solution Technical Information:NaCl (100mM) Tris-HCl, pH 8.0 (10mM) EDTA, pH 8.0 (1mM)
Catalog No. 50-196-670 Supplier Bioworld Supplier No. 105300102
STE Reagent Common reagent for molecular biology. Used in DNA extraction, DNA electrophoresis and many other lab protocols.DNAse and RNAse free, ready to use 1X solution Technical Information:NaCl (100mM) Tris-HCl, pH 8.0 (10mM) EDTA, pH 8.0 (1mM)