Learn More
Bioworld STE Buffer (10X), 4 L
Catalog No. 50196680
Encompass_Preferred
STE ReagentCommon reagent for molecular biology. Used in DNA extraction, DNA electrophoresis and many other lab protocols.DNAse and RNAse free, 10X solutionTechnical Information:1X concentration solution containsNaCl (100mM) Tris-HCl, pH 8.0 (10mM) EDTA, pH 8.0 (1mM)
Catalog No. 50-196-680 Supplier Bioworld Supplier No. 105300152
STE ReagentCommon reagent for molecular biology. Used in DNA extraction, DNA electrophoresis and many other lab protocols.DNAse and RNAse free, 10X solutionTechnical Information:1X concentration solution containsNaCl (100mM) Tris-HCl, pH 8.0 (10mM) EDTA, pH 8.0 (1mM)