Learn More
Research Products International Corp STET Buffer 1X Solution, 500 ML
SDP
Catalog No. 501978369
Encompass_Preferred
Formulation:
NaCI: 100 mM
TRIS HCI: 10 mM
EDTA: 1 mM
Triton X-100: 5%
Catalog No. 50-197-8369 Supplier Research Products International Corp Supplier No. S24300500.0
Formulation:
NaCI: 100 mM
TRIS HCI: 10 mM
EDTA: 1 mM
Triton X-100: 5%