Learn More
Med Vet International Clinician Stethoscope Adscope 603 Black 1-Tube 22 inch Tube Double Sided Chestpiece, 603Bk, Each
Catalog No. 502829828
Encompass_Preferred
Combination diaphragm/bell adult chest piece precisely machined to exacting tolerances from surgical stainless steel for outstanding performance and rugged durability Extra large bell (1-3/8-inch diameter) for unsurpassed low frequency response Ultra-sensitive diaphragm (1-3/4-inch diameter) for greater amplification and crisper high frequency response Non-chill bell and diaphragm retaining rims for patient comfort Clinician headset boasts reinforcing yoke molded into the flexible 22-inch PVC tubing and stainless steel binaurals fixed at 15º angle for greater comfort Adsoft™ threaded PVC ear tips for the ultimate in wearing comfort and acoustic seal
Catalog No. 50-282-9828 Supplier Med Vet International Supplier No. 363826EA
Combination diaphragm/bell adult chest piece precisely machined to exacting tolerances from surgical stainless steel for outstanding performance and rugged durability Extra large bell (1-3/8-inch diameter) for unsurpassed low frequency response Ultra-sensitive diaphragm (1-3/4-inch diameter) for greater amplification and crisper high frequency response Non-chill bell and diaphragm retaining rims for patient comfort Clinician headset boasts reinforcing yoke molded into the flexible 22-inch PVC tubing and stainless steel binaurals fixed at 15º angle for greater comfort Adsoft™ threaded PVC ear tips for the ultimate in wearing comfort and acoustic seal