Learn More
Med Vet International Cardiology Stethoscope 3M Littmann Master Cardiology Purple 1-Tube 27 inch Tube Single Sided Chestpiece, 2167, Each
Catalog No. 502831095
Encompass_Preferred
27 inch tube, Plum Outstanding acoustics: The highest in Littmann mechanical stethoscopes Tunable diaphragm: Hear high or low frequency sounds by slightly adjusting pressure on the chest piece Dual-lumen tubing: Two sound paths in one tube eliminates the rubbing noise of traditional double tubes Headset is easily adjusted for individual fit and comfort. Angled ear tubes align with ear canals Includes Special Procedures Adaptor for pediatric or neonatal auscultation
Catalog No. 50-283-1095 Supplier Med Vet International Supplier No. 520217EA
27 inch tube, Plum Outstanding acoustics: The highest in Littmann mechanical stethoscopes Tunable diaphragm: Hear high or low frequency sounds by slightly adjusting pressure on the chest piece Dual-lumen tubing: Two sound paths in one tube eliminates the rubbing noise of traditional double tubes Headset is easily adjusted for individual fit and comfort. Angled ear tubes align with ear canals Includes Special Procedures Adaptor for pediatric or neonatal auscultation