Learn More
Med Vet International Disposable Stethoscope Proscope 664 Yellow 1-Tube 22 inch Tube Single Sided Chestpiece, 664Y, 50/Case
Catalog No. 502831077
Encompass_Preferred
The Proscope™ stethoscope features a disposable ABS chest piece with a non-chill rim for patient comfort and hygiene Aluminum binaurals deliver clear sound during vital checks of the heart and lungs The yellow stethoscope is able to undergo EtO sterilization It's made of phthalate and latex-free materials to protect sensitive skin A 32-inch length provides flexibility and reach
Catalog No. 50-283-1077 Supplier Med Vet International Supplier No. 493334CS
The Proscope™ stethoscope features a disposable ABS chest piece with a non-chill rim for patient comfort and hygiene Aluminum binaurals deliver clear sound during vital checks of the heart and lungs The yellow stethoscope is able to undergo EtO sterilization It's made of phthalate and latex-free materials to protect sensitive skin A 32-inch length provides flexibility and reach