Learn More
Med Vet International Disposable Stethoscope Proscope 664 Yellow 1-Tube 22 inch Tube Single Sided Chestpiece, 664Y, Each
Catalog No. 502829725
Encompass_Preferred
The Proscope™ stethoscope features a disposable ABS chest piece with a non-chill rim for patient comfort and hygiene Aluminum binaurals deliver clear sound during vital checks of the heart and lungs The yellow stethoscope is able to undergo EtO sterilization It's made of phthalate and latex-free materials to protect sensitive skin A 32-inch length provides flexibility and reach
Catalog No. 50-282-9725 Supplier Med Vet International Supplier No. 493334EA
The Proscope™ stethoscope features a disposable ABS chest piece with a non-chill rim for patient comfort and hygiene Aluminum binaurals deliver clear sound during vital checks of the heart and lungs The yellow stethoscope is able to undergo EtO sterilization It's made of phthalate and latex-free materials to protect sensitive skin A 32-inch length provides flexibility and reach