Learn More
Med Vet International General Exam Stethoscope 3M Littmann Lightweight Ii S.E. Black 1-Tube 28 inch Tube Double Sided Chestpiece, 2450, Each
Catalog No. 502830914
Encompass_Preferred
The 3M™ Littmann lightweight II S. E. stethoscope is lightweight, making it ideal for all-day use A tunable diaphragm provides time-saving convenience The headset tension is easily adjustable to provide a comfortable fit Its resilient 28-inch tubing in black can retain shape quickly The chest piece has an ovoid shape to enhance performance around body contours This double-sided stethoscope features a non-chill rim for enhanced comfort It doesn't contain natural rubber latex, making it safe to use for those with allergies
Catalog No. 50-283-0914 Supplier Med Vet International Supplier No. 372943EA
The 3M™ Littmann lightweight II S. E. stethoscope is lightweight, making it ideal for all-day use A tunable diaphragm provides time-saving convenience The headset tension is easily adjustable to provide a comfortable fit Its resilient 28-inch tubing in black can retain shape quickly The chest piece has an ovoid shape to enhance performance around body contours This double-sided stethoscope features a non-chill rim for enhanced comfort It doesn't contain natural rubber latex, making it safe to use for those with allergies