Learn More
Med Vet International Clinician Stethoscope 3M Littmann Classic Iii Gray 1-Tube 27 inch Tube Convertible Chestpiece, 5621, Each
Catalog No. 502830635
Encompass_Preferred
The double-sided chest piece is easy to grip and move around Adjust the pressure of the 3M™ Classic III™ stethoscope diaphragm to hear either high- or low-frequency sounds You can squeeze or pull apart the ear tubes to adjust the headset The 3M™ Littmann Classic III™ stethoscope has ear tips that conform to the wearer's ears An acoustic seal helps keep out ambient noise The next-generation tubing makes this product durable and helps it retain its shape
Catalog No. 50-283-0635 Supplier Med Vet International Supplier No. 980121EA
The double-sided chest piece is easy to grip and move around Adjust the pressure of the 3M™ Classic III™ stethoscope diaphragm to hear either high- or low-frequency sounds You can squeeze or pull apart the ear tubes to adjust the headset The 3M™ Littmann Classic III™ stethoscope has ear tips that conform to the wearer's ears An acoustic seal helps keep out ambient noise The next-generation tubing makes this product durable and helps it retain its shape