Learn More
Med Vet International Clinician Stethoscope 3M Littmann Classic Iii Purple 1-Tube 27 inch Tube Convertible Chestpiece, 5831, Each
Catalog No. 502830598
Encompass_Preferred
The 27-inch double-sided chest piece stethoscope is easy to grip and move around An acoustic seal helps keep out ambient noise Adjust the pressure of the Littmann Classic III™ stethoscope in plum, 5831 to hear either high- or low-frequency sounds You can squeeze or pull apart the ear tubes to adjust the headset The 3M Classic III™ stethoscope has soft-sealing ear tips that conform to the wearer's ears The next-generation tubing makes this product durable and helps it retain its shape
Catalog No. 50-283-0598 Supplier Med Vet International Supplier No. 995845EA
The 27-inch double-sided chest piece stethoscope is easy to grip and move around An acoustic seal helps keep out ambient noise Adjust the pressure of the Littmann Classic III™ stethoscope in plum, 5831 to hear either high- or low-frequency sounds You can squeeze or pull apart the ear tubes to adjust the headset The 3M Classic III™ stethoscope has soft-sealing ear tips that conform to the wearer's ears The next-generation tubing makes this product durable and helps it retain its shape