Learn More
Med Vet International Clinician Stethoscope 3M Littmann Classic Iii Black 1-Tube 27 inch Tube Convertible Chestpiece, 5620, Each
Catalog No. 502829349
Encompass_Preferred
The 27-inch Littmann 5620 Classic III™ stethoscope is easy to grip and move around An acoustic seal helps keep out ambient noise Adjust the pressure of the Littmann Classic III™ stethoscope in black to hear either high- or low-frequency sounds You can squeeze or pull apart the ear tubes to adjust the headset The 3M Littmann Classic III™ stethoscope has soft-sealing ear tips that conform to the wearer's ears The next-generation tubing makes this product durable and helps it retain its shape
Catalog No. 50-282-9349 Supplier Med Vet International Supplier No. 957763EA
The 27-inch Littmann 5620 Classic III™ stethoscope is easy to grip and move around An acoustic seal helps keep out ambient noise Adjust the pressure of the Littmann Classic III™ stethoscope in black to hear either high- or low-frequency sounds You can squeeze or pull apart the ear tubes to adjust the headset The 3M Littmann Classic III™ stethoscope has soft-sealing ear tips that conform to the wearer's ears The next-generation tubing makes this product durable and helps it retain its shape