Learn More
Med Vet International General Exam Stethoscope 3M Littmann Lightweight Ii S.E. Pink 1-Tube 28 inch Tube Double Sided Chestpiece, 2456, Each
Catalog No. 502831227
Encompass_Preferred
It's the entry-level stethoscope specially designed for basic blood pressure monitoring and physical assessment Double-sided chest piece with unique, easy-access shape Pressure-based sound frequency adjustment with tunable diaphragm Tear-drop shape for easy fit under blood pressure cuff Anatomically designed headset
Catalog No. 50-283-1227 Supplier Med Vet International Supplier No. 846965EA
It's the entry-level stethoscope specially designed for basic blood pressure monitoring and physical assessment Double-sided chest piece with unique, easy-access shape Pressure-based sound frequency adjustment with tunable diaphragm Tear-drop shape for easy fit under blood pressure cuff Anatomically designed headset