Learn More
Med Vet International Cardiology Stethoscope 3M Littmann Cardiology IV Black 1-Tube 22 inch Tube Convertible Chestpiece, 6151, Each
Catalog No. 502829761
Encompass_Preferred
Black Single-piece tunable diaphragm allows better audibility of high frequency sounds Tunable diaphragms on both the adult and pediatric sides of the chest piece Open bell versatility allows the pediatric side to convert to a traditional open bell by replacing the single-piece diaphragm with a non-chill sleeve
Catalog No. 50-282-9761 Supplier Med Vet International Supplier No. 1012629EA
Black Single-piece tunable diaphragm allows better audibility of high frequency sounds Tunable diaphragms on both the adult and pediatric sides of the chest piece Open bell versatility allows the pediatric side to convert to a traditional open bell by replacing the single-piece diaphragm with a non-chill sleeve