Learn More
Med Vet International 3M Littmann Cardiology IV Stethoscope, 27 inch, Black, 6152, Each
Catalog No. 502832157
Encompass_Preferred
Cardiology Stethoscope 3M™ Littmann Cardiology IV™ Black 1-Tube 27 Inch Tube Double-Sided Chestpiece The 3M™ Littmann Cardiology IV™ diagnostic stethoscope is suitable for checking vitals of kids and adults A dual-sided and tunable stainless steel chest piece picks up heartbeats and lung activity in low and high frequencies The bell sleeve prevents chill on the skin from chest piece contact This black Littmann stethoscope has soft-sealing ear tips that stay snugly in place for clear acoustics Ergonomic, high profile construction makes the stethoscope highly flexible and easy to use The black rubber coating is free of latex and safe for sensitive skin
Catalog No. 50-283-2157 Supplier Med Vet International Supplier No. 1013785EA
Cardiology Stethoscope 3M™ Littmann Cardiology IV™ Black 1-Tube 27 Inch Tube Double-Sided Chestpiece The 3M™ Littmann Cardiology IV™ diagnostic stethoscope is suitable for checking vitals of kids and adults A dual-sided and tunable stainless steel chest piece picks up heartbeats and lung activity in low and high frequencies The bell sleeve prevents chill on the skin from chest piece contact This black Littmann stethoscope has soft-sealing ear tips that stay snugly in place for clear acoustics Ergonomic, high profile construction makes the stethoscope highly flexible and easy to use The black rubber coating is free of latex and safe for sensitive skin