Learn More
Med Vet International Cardiology Stethoscope 3M Littmann Master Cardiology Black 1-Tube 27 inch Tube Single Sided Chestpiece, 2161, Each
Catalog No. 502832478
Encompass_Preferred
Tunable diaphragm allows the user to conveniently alternate between the low- and high-frequency sounds without turning over the chest piece Patented tunable diaphragm conveniently alternated between the low and high frequency sounds without the need to turn over the chest piece Handcrafted solid stainless steel chest piece for superior acoustics Two-in-one tube design eliminates tube rubbing noise Excellent acoustic seal and comfortable fit with the soft sealing ear tips Overall length 27 inch
Catalog No. 50-283-2478 Supplier Med Vet International Supplier No. 372940EA
Tunable diaphragm allows the user to conveniently alternate between the low- and high-frequency sounds without turning over the chest piece Patented tunable diaphragm conveniently alternated between the low and high frequency sounds without the need to turn over the chest piece Handcrafted solid stainless steel chest piece for superior acoustics Two-in-one tube design eliminates tube rubbing noise Excellent acoustic seal and comfortable fit with the soft sealing ear tips Overall length 27 inch