Learn More
Med Vet International Cardiology Stethoscope 3M Littmann Master Cardiology Black 1-Tube 27 inch Tube Single Sided Chestpiece, 2160, Each
Catalog No. 502830058
Encompass_Preferred
The black 3M™ Littmann Master Cardiology™ stethoscope provides clear acoustics for accurate diagnostics A tunable diaphragm allows for hearing low or high-frequency sounds by simply adjusting chest piece pressure Angled ear tubes aligned with ear canals and an adjustable headset offer a customized, comfortable fit Sealing ear tips stay snugly in place and further improve acoustic quality Attach the included special procedures adapter for neonatal or pediatric applications The 27-inch 3M™ Littmann Master Cardiology™ stethoscope features next-generation tubing that's oil- and alcohol-resistant as well as free of latex
Catalog No. 50-283-0058 Supplier Med Vet International Supplier No. 372939EA
The black 3M™ Littmann Master Cardiology™ stethoscope provides clear acoustics for accurate diagnostics A tunable diaphragm allows for hearing low or high-frequency sounds by simply adjusting chest piece pressure Angled ear tubes aligned with ear canals and an adjustable headset offer a customized, comfortable fit Sealing ear tips stay snugly in place and further improve acoustic quality Attach the included special procedures adapter for neonatal or pediatric applications The 27-inch 3M™ Littmann Master Cardiology™ stethoscope features next-generation tubing that's oil- and alcohol-resistant as well as free of latex