Learn More
Med Vet International General Exam Stethoscope 3M Littmann Lightweight Ii S.E. Burgundy 1-Tube 28 inch Tube Double Sided Chestpiece, 2451, Each
Catalog No. 502832430
Encompass_Preferred
The 3M™ Littmann Lightweight II S. E. Stethoscope is lightweight, making it ideal for all-day use A tunable diaphragm provides time-saving convenience The headset tension is easily adjustable to provide a comfortable fit Its resilient tubing can retain shape quickly The chest piece has an ovoid shape to enhance performance around body contours It features a non-chill rim for enhanced comfort It doesn't contain natural rubber latex, making it safe to use for those with allergies
Catalog No. 50-283-2430 Supplier Med Vet International Supplier No. 466397EA
The 3M™ Littmann Lightweight II S. E. Stethoscope is lightweight, making it ideal for all-day use A tunable diaphragm provides time-saving convenience The headset tension is easily adjustable to provide a comfortable fit Its resilient tubing can retain shape quickly The chest piece has an ovoid shape to enhance performance around body contours It features a non-chill rim for enhanced comfort It doesn't contain natural rubber latex, making it safe to use for those with allergies