Learn More
Med Vet International Miltex Vantage Stevens Tenotomy Scissors, 4.125in, Curved
Catalog No. 502522280
Encompass_Preferred
The Miltex Vantage V918-1466 Stevens Tenotomy Scissors have a length of 4.125in. and arre made of Mid-Grade stainless steel. This Stevens Tenotomy Scissors are used for delicate dissection and cutting and have small, sharp blades, and has a curved tip.
Catalog No. 50-252-2280 Supplier Med Vet International Supplier No. MILV9181466
The Miltex Vantage V918-1466 Stevens Tenotomy Scissors have a length of 4.125in. and arre made of Mid-Grade stainless steel. This Stevens Tenotomy Scissors are used for delicate dissection and cutting and have small, sharp blades, and has a curved tip.