Learn More
Med Vet International Miltex Vantage Stevens Tenotomy Scissors, 4.125in., Straight
Catalog No. 502830414
Encompass_Preferred
The Miltex Vantage V918-1462 Stevens Tenotomy Scissors have a length of 4.125in. and are made of Mid-Grade stainless steel. This Stevens Tenotomy Scissors are used for delicate dissection and cutting and have small, sharp blades, and has a straight tip.
Catalog No. 50-283-0414 Supplier Med Vet International Supplier No. MILV9181462
The Miltex Vantage V918-1462 Stevens Tenotomy Scissors have a length of 4.125in. and are made of Mid-Grade stainless steel. This Stevens Tenotomy Scissors are used for delicate dissection and cutting and have small, sharp blades, and has a straight tip.