Learn More
Medchemexpress LLC Steviol | 471-80-7 | 318.45 | 5 MG
SDP
Catalog No. 5000415222
Encompass_Preferred
Steviol is the main metabolite of steviol glycosides and an inhibitor of AQP2/CFTR. It slows down renal cyst growth by inhibiting CFTR activity, reducing AQP2 expression, and promoting AQP2 and CFTR degradation. It can be used in polycystic kidney disease research.
  • Main metabolite of steviol glycosides
  • Inhibitor of AQP2/CFTR
  • Slows down the growth of renal cysts
  • Inhibits CFTR activity
  • Reduces AQP2 expression
  • Promotes degradation of AQP2 and CFTR
  • Can be used in polycystic kidney disease research
  • Exerts diuretic, natriuretic, kaliuretic effects
  • Inhibits renal tubular reabsorption of glucose
Catalog No. 50-004-15222 Supplier Medchemexpress LLC Supplier No. HYN20575MG
Steviol is the main metabolite of steviol glycosides and an inhibitor of AQP2/CFTR. It slows down renal cyst growth by inhibiting CFTR activity, reducing AQP2 expression, and promoting AQP2 and CFTR degradation. It can be used in polycystic kidney disease research.
  • Main metabolite of steviol glycosides
  • Inhibitor of AQP2/CFTR
  • Slows down the growth of renal cysts
  • Inhibits CFTR activity
  • Reduces AQP2 expression
  • Promotes degradation of AQP2 and CFTR
  • Can be used in polycystic kidney disease research
  • Exerts diuretic, natriuretic, kaliuretic effects
  • Inhibits renal tubular reabsorption of glucose