Learn More
Cayman Chemical Steviolbioside, Purity: ≥98%, CAS: 41093-60-1, 50 mg
SDP
Catalog No. 502912319
Encompass_Preferred
A natural non-caloric sweetener and an active metabolite of rebaudioside A; inhibits the proliferation of MDA-MB-231 cells at 250 µg/ml; active against M. tuberculosis (MIC = 3.75 µg/ml)
Catalog No. 50-291-2319 Supplier Cayman Chemical Supplier No. 3651350 MG
A natural non-caloric sweetener and an active metabolite of rebaudioside A; inhibits the proliferation of MDA-MB-231 cells at 250 µg/ml; active against M. tuberculosis (MIC = 3.75 µg/ml)