Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stevioside

Catalog No. AC461560250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461560250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC461560250 Supplier Thermo Scientific Chemicals Supplier No. 461560250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57817-89-7 | C38H60O18 | 804.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57817-89-7
Molecular Formula C38H60O18
Molecular Weight (g/mol) 804.88
InChI Key UEDUENGHJMELGK-PEKONUEISA-N
Synonym stevioside
PubChem CID 134129657
SMILES CC12CCCC(C1CCC34C2CCC(C3)(C(=C)C4)OC5C(C(C(C(O5)CO)O)O)OC6C(C(C(C(O6)CO)O)O)O)(C)C(=O)OC7C(C(C(C(O7)CO)O)O)O

Specifications

Melting Point 198.0°C
Loss on Drying 6% max.
Infrared Spectrum Authentic
Quantity 25 g
Solubility Information Solubility in water: 1,25g/L.
Formula Weight 804.88
Percent Purity ≥75% (HPLC)
Chemical Name or Material Stevioside
EINECSNumber 260-975-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.