Learn More
Cayman Chemical Stevioside (hydrate), Purity: ≥95%, 50 mg
SDP
Catalog No. 502954303
Encompass_Preferred
A natural non-caloric sweetener; increases glucose-stimulated insulin secretion in isolated mouse pancreatic islets at 1 µM; reduces LPS-stimulated increases in TNF-α and IL-1β production in THP-1 cells, but also induces TNF-α and IL-1β production in unstimulated THP-1 cells at 1 mM; decreases systolic blood pressure in GK type 2 diabetic rats at 0.03 g/kg; reduces TPA-induced ear edema in mice (ID50 = 291.6 µg/ear)
Catalog No. 50-295-4303 Supplier Cayman Chemical Supplier No. 1190250 MG
A natural non-caloric sweetener; increases glucose-stimulated insulin secretion in isolated mouse pancreatic islets at 1 µM; reduces LPS-stimulated increases in TNF-α and IL-1β production in THP-1 cells, but also induces TNF-α and IL-1β production in unstimulated THP-1 cells at 1 mM; decreases systolic blood pressure in GK type 2 diabetic rats at 0.03 g/kg; reduces TPA-induced ear edema in mice (ID50 = 291.6 µg/ear)