Learn More
Selleck Chemical LLC STF-31
SDP
Catalog No. 5000001107
Encompass_Preferred
STF-31 is a selective glucose transporter GLUT1 inhibitor
Catalog No. 50-000-01107 Supplier Selleck Chemical LLC Supplier No. S793110MM/1ML
STF-31 is a selective glucose transporter GLUT1 inhibitor