Learn More
Apexbio Technology LLC STF 31 724741-75-7 50mg
SDP
Catalog No. 5000091753
Encompass_Preferred
STF 31 (CAS 724741-75-7) is a small-molecule inhibitor targeting glucose transporter 1 (GLUT1) with an IC50 of 1 M By antagonizing GLUT1 STF 31 disrupts glucose uptake selectively impairing survival in VHL-deficient renal cell carcinoma (RCC) models that depend on enhanced glycolysis In vitro STF 31 exhibits selective cytotoxicity toward VHL-deficient RCC4 cells while exerting minimal effects on RCC4/VHL controls In vivo administration of STF 31 analogs retards tumor growth in VHL-deficient RCC xenografts without notable toxicity in mice STF 31 is utilized in research to investigate GLUT1-mediated metabolic dependencies and therapeutic vulnerabilities in cancer and neuroinflammatory models
Catalog No. 50-000-91753 Supplier Apexbio Technology LLC Supplier No. B767050
STF 31 (CAS 724741-75-7) is a small-molecule inhibitor targeting glucose transporter 1 (GLUT1) with an IC50 of 1 M By antagonizing GLUT1 STF 31 disrupts glucose uptake selectively impairing survival in VHL-deficient renal cell carcinoma (RCC) models that depend on enhanced glycolysis In vitro STF 31 exhibits selective cytotoxicity toward VHL-deficient RCC4 cells while exerting minimal effects on RCC4/VHL controls In vivo administration of STF 31 analogs retards tumor growth in VHL-deficient RCC xenografts without notable toxicity in mice STF 31 is utilized in research to investigate GLUT1-mediated metabolic dependencies and therapeutic vulnerabilities in cancer and neuroinflammatory models