Learn More
Medchemexpress LLC STF-31 | 724741-75-7 | 97.0% | 423.53 | 10 MM 1 ML
SDP
Catalog No. 5000385513
Encompass_Preferred
STF-31 is a selective inhibitor of glucose transporter 1 (GLUT1) with an IC50 of 1 μM. It also functions as a NAMPT inhibitor and inhibits glucose uptake in renal cell carcinoma (RCC) 4 cells.
  • Targets GLUT
  • Targets Autophagy
  • Used in cancer research
  • Cited in over 15 publications
Catalog No. 50-003-85513 Supplier Medchemexpress LLC Supplier No. HY1872810MM/1ML
STF-31 is a selective inhibitor of glucose transporter 1 (GLUT1) with an IC50 of 1 μM. It also functions as a NAMPT inhibitor and inhibits glucose uptake in renal cell carcinoma (RCC) 4 cells.
  • Targets GLUT
  • Targets Autophagy
  • Used in cancer research
  • Cited in over 15 publications