Learn More
AdipoGen STF-31
SDP
Catalog No. 502562212
Encompass_Preferred
Chemical. CAS 724741-75-7. Formula C23H25N3O3S. MW 423.5. . Specific nicotinamide phosphoribosyltransferase NAMPT inhibitor IC50 = 19nM, consequently blocking the NAD⁺ salvage pathway and leading to blockage of GLUT1 expression. Glucose transporter 1 GLUT1 inhibitor IC50 ~1µM. Inhibits glucose uptake across the plasma membranes of mammalian cells and consequently decreases glycolysis. Anticancer agent. Shown to induce necrosis in cancer cells that lack the von Hippel-Lindau VHL tumor suppressor gene, which overexpress GLUT1. VHL-deficient cancer cells are dependent on the high affinity GLUT1 transporter and aerobic glycolysis for ATP production. Completely suppressed the glucose uptake activity and induced apoptosis in GLUT1 expressing myeloma cells as well. Useful agent for immunometabolism research. Toxic to human pluripotent stem cells hPSCs and used to selectively eliminate hPSCs from mixed cultures.
Catalog No. 50-256-2212 Supplier AdipoGen Supplier No. AGCR13693M025
Chemical. CAS 724741-75-7. Formula C23H25N3O3S. MW 423.5. . Specific nicotinamide phosphoribosyltransferase NAMPT inhibitor IC50 = 19nM, consequently blocking the NAD⁺ salvage pathway and leading to blockage of GLUT1 expression. Glucose transporter 1 GLUT1 inhibitor IC50 ~1µM. Inhibits glucose uptake across the plasma membranes of mammalian cells and consequently decreases glycolysis. Anticancer agent. Shown to induce necrosis in cancer cells that lack the von Hippel-Lindau VHL tumor suppressor gene, which overexpress GLUT1. VHL-deficient cancer cells are dependent on the high affinity GLUT1 transporter and aerobic glycolysis for ATP production. Completely suppressed the glucose uptake activity and induced apoptosis in GLUT1 expressing myeloma cells as well. Useful agent for immunometabolism research. Toxic to human pluripotent stem cells hPSCs and used to selectively eliminate hPSCs from mixed cultures.