Learn More
Apexbio Technology LLC STF-62247, 10mg. Cas: 315702-99-9 MFCD: MFCD01121974. Autophagy inducer in renal cell
SDP
Supplier:  Apexbio Technology LLC B215110
Encompass_Preferred
STF-62247 shows selectivtoxicity and growth inhibition of renal cells lacking VHL; 25-fold greater sensitivity observed for cells with VHL deficiency compared to wild-type (VHL+). 315702-99-9. MFCD01121974
Catalog No. 50-101-2042