Learn More
Lonza Walkersville StGM Stellate Cell Culture BulletKit
Catalog No. NC2221989
Encompass
Bullet Kit consisting of basal medium plus supplements for culturing human liver stellate cells.For use with Stellate cell products HUCLS-200K and HUCLS-1M.For HUCLS and HUCLS1, please use MCST250.Content:500mL Basal MediumSingleQuot Supplements
This item is not returnable. View return policy
Catalog No. NC2221989 Supplier Lonza Walkersville Supplier No. MST500BK
Bullet Kit consisting of basal medium plus supplements for culturing human liver stellate cells.For use with Stellate cell products HUCLS-200K and HUCLS-1M.For HUCLS and HUCLS1, please use MCST250.Content:500mL Basal MediumSingleQuot Supplements