Learn More
Medchemexpress LLC HY-P2695 1mg Medchemexpress, STh CAS:118447-40-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP26951MG
Encompass_Preferred
Medchemexpress, HY-P2695 1mg STh CAS:118447-40-8 STh, an Escherichia coli heat-stable toxin, is a 19 amino acid polypeptide encompassing three disulfide bridges. STh is an antigen of interest in the search for a broad coverage enterotoxigenic Escherichia coli (ETEC) vaccine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3091