Learn More
Accuflow Systems STHT-C Tubing 50-foot, ID 0.250"" x OD 0.375"" x Wall 0.063"" is ultra-pure biopharmaceutical-grade silicone with unsurpassed surface smoothness. STHT-C is autoclavable and sterilizable, has a temperature range from 80F (-62C) to 500F (260C)
Catalog No. 501539558
Encompass_Preferred
Sani-Tech STHT-C is an ultra-pure biopharmaceutical-grade tubing. This platinum-cured silicone tubing resists temperature extremes, ozone, radiation, moisture, compression sets, weathering and chemical attack, and imparts no odors to fluids transported within it. Sani-Tech STHT-C silicone tubing is suitable for sterilization by gamma irradiation and steam. Sani-Tech STHT-C, in 1/16'' ID and greater, comes with laser-marked part numbers and lot codes to provide total product traceability. Meets USP Class VI, European Pharmacopoeia 3.1.9 and FDA criteria.
Catalog No. 50-153-9558 Supplier Accuflow Systems Supplier No. STHTC2502
Sani-Tech STHT-C is an ultra-pure biopharmaceutical-grade tubing. This platinum-cured silicone tubing resists temperature extremes, ozone, radiation, moisture, compression sets, weathering and chemical attack, and imparts no odors to fluids transported within it. Sani-Tech STHT-C silicone tubing is suitable for sterilization by gamma irradiation and steam. Sani-Tech STHT-C, in 1/16'' ID and greater, comes with laser-marked part numbers and lot codes to provide total product traceability. Meets USP Class VI, European Pharmacopoeia 3.1.9 and FDA criteria.