Learn More
Accuflow Systems Sani-Tech STHT-R tubing, 50-foot, ID 0.500"" x OD 0.875"" x Wall 0.187"" is manufactured from the finest grade of silicone materials and is fully characterized, validated and tested to a variety of specifications including USP Class VI criteria, ISO 10993 and European Pharmacopoeia 3.1.9.
Catalog No. 501539405
Encompass_Preferred
Sani-Tech STHT-R platinum-cured braid-reinforced silicone hose is an ultra-flexible, high-purity hose that was developed for higher pressure applications. STHT-R is manufactured with Sani-Tech 65 brand silicone resin. Sani-Tech STHT-R platinum-cured silicone hose resists temperature extremes, ozone, corona, radiation, moisture, compression sets, weathering and chemical attack and imparts no taste or odors to fluids transported within it. Sani-Tech STHT-R hose withstands repeated autoclaving and resists the adherence of bloodproducts and other sanitary fluids.
Catalog No. 50-153-9405 Supplier Accuflow Systems Supplier No. STHTR0500
Sani-Tech STHT-R platinum-cured braid-reinforced silicone hose is an ultra-flexible, high-purity hose that was developed for higher pressure applications. STHT-R is manufactured with Sani-Tech 65 brand silicone resin. Sani-Tech STHT-R platinum-cured silicone hose resists temperature extremes, ozone, corona, radiation, moisture, compression sets, weathering and chemical attack and imparts no taste or odors to fluids transported within it. Sani-Tech STHT-R hose withstands repeated autoclaving and resists the adherence of bloodproducts and other sanitary fluids.