Learn More
MSC Gf Gage Thread Insert Plug Gage, 5/16-18, Steel, 2B, No Go, 1 ea
Catalog No. 5000512097
Encompass
This steel screw thread insert (STI) plug thread No Go gage is designed for inspecting internal threads after the installation of screw thread inserts. It provides a precise method for ensuring that the hole size and thread profile meet the required 2B class specifications.
  • Constructed from durable steel for long-lasting use in shop environments
  • Designed for 5/16-18 thread sizes
  • Single-end configuration
  • Class 2B tolerance for reliable inspection
Catalog No. 50-005-12097 Supplier MSC Supplier No. 55315279
This steel screw thread insert (STI) plug thread No Go gage is designed for inspecting internal threads after the installation of screw thread inserts. It provides a precise method for ensuring that the hole size and thread profile meet the required 2B class specifications.
  • Constructed from durable steel for long-lasting use in shop environments
  • Designed for 5/16-18 thread sizes
  • Single-end configuration
  • Class 2B tolerance for reliable inspection