Learn More
Cayman Chemical STI1BIo-Rad MultIplexed qP 1ea
SDP
Catalog No. 502945657
Encompass_Preferred
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X qPCR enzyme mastermix Organism Type: Bacteria Gram-, Protozoan, Multiplexed Reaction Panels: STI, Vaginal microbiota Colors: Cy5 (Trichomonas vaginalis), Cy5.5 (Mycoplasma hominis), FAM (Chlamydia trachomatis), HEX (RPP30), TEX615 (Haemophilus ducreyi) Instruments: Bio-Rad CFX96 or CFX384
Catalog No. 50-294-5657 Supplier Cayman Chemical Supplier No. 406841 EA
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X qPCR enzyme mastermix Organism Type: Bacteria Gram-, Protozoan, Multiplexed Reaction Panels: STI, Vaginal microbiota Colors: Cy5 (Trichomonas vaginalis), Cy5.5 (Mycoplasma hominis), FAM (Chlamydia trachomatis), HEX (RPP30), TEX615 (Haemophilus ducreyi) Instruments: Bio-Rad CFX96 or CFX384