Learn More
Cayman Chemical STI2 (Bio-Rad) Multiplexed qPCR Assay, 1 ea
SDP
Catalog No. 502942936
Encompass_Preferred
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X qPCR enzyme mastermix Organism Type: Bacteria (no cell well), Multiplexed Reaction Panels: STI, Vaginal microbiota Colors: Cy5 (Mycoplasma genitalium), Cy5.5 (Ureaplasma urealyticum), FAM (Neisseria gonorrhoeae), HEX (RPP30), TEX615 (Treponema pallidum) Instruments: Bio-Rad CFX96 or CFX384
Catalog No. 50-294-2936 Supplier Cayman Chemical Supplier No. 406851 EA
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X qPCR enzyme mastermix Organism Type: Bacteria (no cell well), Multiplexed Reaction Panels: STI, Vaginal microbiota Colors: Cy5 (Mycoplasma genitalium), Cy5.5 (Ureaplasma urealyticum), FAM (Neisseria gonorrhoeae), HEX (RPP30), TEX615 (Treponema pallidum) Instruments: Bio-Rad CFX96 or CFX384