Learn More
Cayman Chemical STI3 (Bio-Rad) Multiplexed qPCR Assay Bundle, 1 ea
SDP
Catalog No. 502944010
Encompass_Preferred
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Kit Includes: Tube 1: 5X Primer/Probe mix (5X) for C. trachomatis, N. gonorrhoeae, T. pallidum, HSV1, HSV2, and hRPP30DNA Tube 2: Positive control: 5000 copies/µl positive controls of synthetic 500 bp DNA fragments of C. trachomatis, N. gonorrhoeae, T. pallidum, HSV1, HSV2, and hRPP30DNA Tube 3: Standard qPCR enzyme mastermix (Item No. 40706) Panels: STI, Vaginal microbiota Colors: Cy5 (Chlamydia trachomatis), Cy5.5 (Neisseria gonorrhoeae), FAM (HSV-1 and HSV-2), HEX (RPP30), TEX615 (Treponema pallidum) Instruments: Bio-Rad CFX96 or CFX384
Catalog No. 50-294-4010 Supplier Cayman Chemical Supplier No. 406861 EA
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Kit Includes: Tube 1: 5X Primer/Probe mix (5X) for C. trachomatis, N. gonorrhoeae, T. pallidum, HSV1, HSV2, and hRPP30DNA Tube 2: Positive control: 5000 copies/µl positive controls of synthetic 500 bp DNA fragments of C. trachomatis, N. gonorrhoeae, T. pallidum, HSV1, HSV2, and hRPP30DNA Tube 3: Standard qPCR enzyme mastermix (Item No. 40706) Panels: STI, Vaginal microbiota Colors: Cy5 (Chlamydia trachomatis), Cy5.5 (Neisseria gonorrhoeae), FAM (HSV-1 and HSV-2), HEX (RPP30), TEX615 (Treponema pallidum) Instruments: Bio-Rad CFX96 or CFX384