Learn More
Cayman Chemical STI4 (Bio-Rad) Multiplexed qPCR Assay, 1 ea
SDP
Catalog No. 502943263
Encompass_Preferred
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X qPCR enzyme mastermix Organism Type: Bacteria Gram-, Virus dsDNA, Multiplexed Reaction Panels: STI, Vaginal microbiota Colors: Cy5 (Chlamydia trachomatis), FAM (HSV-1 and HSV-2), HEX (RPP30), TEX615 (Neisseria gonorrhoeae) Instruments: Bio-Rad CFX96 or CFX384
Catalog No. 50-294-3263 Supplier Cayman Chemical Supplier No. 406871 EA
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X qPCR enzyme mastermix Organism Type: Bacteria Gram-, Virus dsDNA, Multiplexed Reaction Panels: STI, Vaginal microbiota Colors: Cy5 (Chlamydia trachomatis), FAM (HSV-1 and HSV-2), HEX (RPP30), TEX615 (Neisseria gonorrhoeae) Instruments: Bio-Rad CFX96 or CFX384