Learn More
Cayman Chemical STI6LesIonsBIo-Rad Mult 1ea
SDP
Catalog No. 502945556
Encompass_Preferred
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X qPCR enzyme mastermix Organism Type: Bacteria Gram-, Virus dsDN,, Multiplexed Reaction Panels: STI, Vaginal microbiota Colors: Cy5 (HSV-1), Cy5.5 (HSV-2), FAM (MPX), HEX (RPP30), TEX615 (Treponema pallidum) Instruments: Bio-Rad CFX96 or CFX384
Catalog No. 50-294-5556 Supplier Cayman Chemical Supplier No. 406891 EA
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X qPCR enzyme mastermix Organism Type: Bacteria Gram-, Virus dsDN,, Multiplexed Reaction Panels: STI, Vaginal microbiota Colors: Cy5 (HSV-1), Cy5.5 (HSV-2), FAM (MPX), HEX (RPP30), TEX615 (Treponema pallidum) Instruments: Bio-Rad CFX96 or CFX384