Learn More
Cayman Chemical STI7 (general STI often found together) (Bio-Rad) Multiplexed qPCR Assay Bundle, 1 ea
SDP
Catalog No. 502945557
Encompass_Preferred
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X qPCR enzyme mastermix Organism Type: Bacteria (no cell wall), Bacteria Gram-, Protozoan, Multiplexed Reaction Panels: STI, Vaginal microbiota Colors: Cy5 (Mycoplasma genitalium), Cy5.5 (Neisseria gonorrhoeae), FAM (Chlamydia trachomatis), HEX (RPP30), TEX615 Instruments: Bio-Rad CFX96 or CFX384 (Trichomonas vaginalis)
Catalog No. 50-294-5557 Supplier Cayman Chemical Supplier No. 406901 EA
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X qPCR enzyme mastermix Organism Type: Bacteria (no cell wall), Bacteria Gram-, Protozoan, Multiplexed Reaction Panels: STI, Vaginal microbiota Colors: Cy5 (Mycoplasma genitalium), Cy5.5 (Neisseria gonorrhoeae), FAM (Chlamydia trachomatis), HEX (RPP30), TEX615 Instruments: Bio-Rad CFX96 or CFX384 (Trichomonas vaginalis)