Learn More
Bachem Stichodactyla helianthus Neurotoxin (ShK) Trifluoroacetate
SDP
Supplier:  Bachem 4031286.0500
Encompass_Preferred
Stichodactyla helianthus Neurotoxin (ShK) Trifluoroacetate, H-Arg-Ser-Cys-Ile-Asp-Thr-Ile-Pro-Lys-Ser-Arg-Cys-Thr-Ala-Phe-Gln-Cys-Lys-His-Ser-Met-Lys-Tyr-Arg-Leu-Ser-Phe-Cys-Arg-Lys-Thr-Cys-Gly-Thr-Cys-OH, 172450-46-3, C169H274N54O48S7, Mr 4054.83, 0.5mg. ShK. ShK-toxin, originally isolated from the sea anemone Stichodactyla helianthus, has been found to inhibit the specific binding of dendrotoxin I to rat brain membranes. Due to its unique structure (it contains three intramolecular disulfide bridges) and high affinity for some potassium channels, ShK-toxin may become a useful molecular probe for investigating potassium channels.
Catalog No. 50-259-594