Learn More
Carbosynth LLC. Stichodactyla Toxin (ShK) (Amide), 1mg, MW: (M.W. 4053.86), Molecular formula: C169H275N55O47S7
SDP
Catalog No. 501687118
Encompass_Preferred
Synonyms: H-Arg-Ser-Cys-Ile-Asp-Thr-Ile-Pro-Lys-Ser-Arg-Cys-Thr-Ala-Phe-Gln-Cys-Lys-His-Ser-Met-Lys-Tyr-Arg-Leu-Ser-Phe-Cys-Arg-Lys-Thr-Cys-Gly-Thr-Cys-NH2, Sea Anemone, Stichodactyla helianthus, Application: Voltage Dependent K+ Channel (A Channel) Blocker, Storage Temperature: -20°C, References: E. Karlsson, et al., Toxicon, 31, 504 (1993). (Original; in Abstract), O. Castañeda, et al., Toxicon, 33, 603 (1995). (Pharmacol.), B. Dang, et al., J Biol Chem, (2017).(Epub ahead of print) doi: 10.1074/jbc.M117.793943
Catalog No. 50-168-7118 Supplier Carbosynth LLC. Supplier No. SHK3770PI1MG
Synonyms: H-Arg-Ser-Cys-Ile-Asp-Thr-Ile-Pro-Lys-Ser-Arg-Cys-Thr-Ala-Phe-Gln-Cys-Lys-His-Ser-Met-Lys-Tyr-Arg-Leu-Ser-Phe-Cys-Arg-Lys-Thr-Cys-Gly-Thr-Cys-NH2, Sea Anemone, Stichodactyla helianthus, Application: Voltage Dependent K+ Channel (A Channel) Blocker, Storage Temperature: -20°C, References: E. Karlsson, et al., Toxicon, 31, 504 (1993). (Original; in Abstract), O. Castañeda, et al., Toxicon, 33, 603 (1995). (Pharmacol.), B. Dang, et al., J Biol Chem, (2017).(Epub ahead of print) doi: 10.1074/jbc.M117.793943